Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monocarboxylic Acid Transporter 1 Antibody (YA3244) | 10 UL
SDP

Catalog No. 5000445198
Encompass_Preferred

Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Monocarboxylic Acid Transporter 1. This protein acts as a proton-linked monocarboxylate transporter, crucial for moving substances such as lactate and pyruvate across cell membranes. Mutations in the gene associated with this transporter can lead to erythrocyte lactate transporter defects.

  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Targets monocarboxylic acid transporter 1
  • Reactivity: Human
  • Molecular weight: 54 kDa
  • Purity: Affinity purified
  • Application: WB, with a dilution ratio of 1:500-1:1000
  • Formulation includes 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Appearance: Liquid

Catalog No. 50-004-45198 Supplier Medchemexpress LLC Supplier No. HYP8349910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Monocarboxylic Acid Transporter 1. This protein acts as a proton-linked monocarboxylate transporter, crucial for moving substances such as lactate and pyruvate across cell membranes. Mutations in the gene associated with this transporter can lead to erythrocyte lactate transporter defects.

  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Targets monocarboxylic acid transporter 1
  • Reactivity: Human
  • Molecular weight: 54 kDa
  • Purity: Affinity purified
  • Application: WB, with a dilution ratio of 1:500-1:1000
  • Formulation includes 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Appearance: Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.