Learn More
Medchemexpress LLC Monocarboxylic Acid Transporter 1 Antibody (YA3244) | 100 UL
SDP
Catalog No. 5000445259
Encompass_Preferred
Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Monocarboxylic Acid Transporter 1. This protein facilitates the movement of monocarboxylates like lactate and pyruvate across cell membranes. Mutations in its gene are linked to erythrocyte lactate transporter defect. The product is supplied in a buffered solution and is suitable for Western Blot (WB) applications.
Features and benefits:
  • Rabbit host
  • Human reactivity
  • Non-conjugated
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Liquid appearance
  • Recommended dilution ratio for WB: 1:500-1:1000
Catalog No. 50-004-45259 Supplier Medchemexpress LLC Supplier No. HYP83499100UL
Add to Cart
Add to Cart
Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Monocarboxylic Acid Transporter 1. This protein facilitates the movement of monocarboxylates like lactate and pyruvate across cell membranes. Mutations in its gene are linked to erythrocyte lactate transporter defect. The product is supplied in a buffered solution and is suitable for Western Blot (WB) applications.
Features and benefits:
  • Rabbit host
  • Human reactivity
  • Non-conjugated
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Liquid appearance
  • Recommended dilution ratio for WB: 1:500-1:1000