Learn More
Medchemexpress LLC Monocarboxylic acid transporter 1 antibody (YA3244) | 50 UL
SDP
Catalog No. 5000445154
Encompass_Preferred
Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a rabbit-derived, non-conjugated IgG monoclonal antibody, targeting Monocarboxylic Acid Transporter 1. It catalyzes the movement of monocarboxylates like lactate and pyruvate across the plasma membrane. This product is intended for research use only and should be handled by experienced, qualified scientists.
  • Rabbit-derived IgG monoclonal antibody
  • Targets monocarboxylic acid transporter 1
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Stable under recommended storage conditions
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-45154 Supplier Medchemexpress LLC Supplier No. HYP8349950UL
Monocarboxylic Acid Transporter 1 Antibody (YA3244) is a rabbit-derived, non-conjugated IgG monoclonal antibody, targeting Monocarboxylic Acid Transporter 1. It catalyzes the movement of monocarboxylates like lactate and pyruvate across the plasma membrane. This product is intended for research use only and should be handled by experienced, qualified scientists.
  • Rabbit-derived IgG monoclonal antibody
  • Targets monocarboxylic acid transporter 1
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Stable under recommended storage conditions
  • Suitable for laboratory centrifugation and storage