Learn More
Hach Company Monochlor F Reagent Powder Pillows, pk/50
Supplier:  Hach Company 2802246
Encompass
See DownloadsOtherProduct Notice-Monochloramine and Free Ammonia for details on accuracy improvements.For determination of monochloramine and free ammonia by the Indophenol method. Hach Methods 10171, 10172 and 10200. Pack of 50 powder pillows.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6695