Learn More
Aqua Solutions MONOCHLOROACETIC ACID BUFFER
SDP
Catalog No. NC2228558
Encompass_Preferred
MONOCHLOROACETIC ACID BUFFER pH 2.8 1L Plastic shelf life 2 months.
This item is not returnable. View return policy
Catalog No. NC2228558 Supplier Aqua Solutions Supplier No. DOW742
MONOCHLOROACETIC ACID BUFFER pH 2.8 1L Plastic shelf life 2 months.