Learn More
Abcam Monochlorobimane, 25MG
Catalog No. 502671198
Encompass_Preferred
MW 226.66 Da, Purity >=98%. Monochlorobimane is a cell-permeant probe for quantifying glutathione levels in cells. It is essentially nonfluorescent until it reacts with a thiol group. It readily reacts with several low molecular weight thiols, including cysteine, glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monochlorobimane has an excitation/emission maxima ~394/490 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1198 Supplier Abcam Supplier No. AB28712925MG
MW 226.66 Da, Purity >=98%. Monochlorobimane is a cell-permeant probe for quantifying glutathione levels in cells. It is essentially nonfluorescent until it reacts with a thiol group. It readily reacts with several low molecular weight thiols, including cysteine, glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monochlorobimane has an excitation/emission maxima ~394/490 nm.
The product is subject to the following: Abcam Restricted Use Statement