Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monochlorobimane (mBCI)

Catalog No. M1381MP
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1381MP 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1381MP Supplier Invitrogen™ Supplier No. M1381MP
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

monochlorobimane (mBCI)

Monochloromobimane is essentially nonfluorescent until conjugated, readily reacts with several low molecular weight thiols, including glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monochlorobimane has absorption/emission maxima ∼394/490 nm.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Monochlorobimane
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Room Temperature
Emission 490

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.