Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti-IdU antibody produced in mouse, 100 ug
Catalog No. NC1794843
Encompass
Anti-IdU Monoclonal Antibody was purified from ascites fluid by Protein A chromatography. This antibody is specific for IdU. Cross-reactivity is not observed with BrdU, CldU or FdU.
Catalog No. NC1794843 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAB3701448100UG
Anti-IdU Monoclonal Antibody was purified from ascites fluid by Protein A chromatography. This antibody is specific for IdU. Cross-reactivity is not observed with BrdU, CldU or FdU.