Learn More
ACROBiosystems ACROBiosystems Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control
Supplier:  ACROBiosystems DNPM1A125TESTS
Encompass_Preferred
Antibody;25TESTS;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.; ;2e5 of anti-CD19 CAR-293 cells were stained with 100ul of 1:50 dilution of Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1A1) and positive control antibody respectively, washed and then followed by PE anti-mouse IgG1 and analyzed with FACS (QC tested).
Catalog No. 50-210-7391