Learn More
ACROBiosystems ACROBiosystems Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Carrier free, MALS verified)
Catalog No. 502106622
Encompass_Preferred
Antibody;100UG;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies.Monoclonal Anti-DNP (Anti-Hapten) antibody, Mouse IgG1 was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins. ;Immobilized DNP-Albumin Conjugate, Bovine at 10 μg/mL (100 μL/well) can bind Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-210-6622 Supplier ACROBiosystems Supplier No. DNPM1100UG
Antibody;100UG;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies.Monoclonal Anti-DNP (Anti-Hapten) antibody, Mouse IgG1 was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins. ;Immobilized DNP-Albumin Conjugate, Bovine at 10 μg/mL (100 μL/well) can bind Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1) with a linear range of 2-20 ng/mL (QC tested).