Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal ANTI-FLAG(R) M2 antibody produced in mouse 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4) | 50UG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F180450UG
Encompass
Monoclonal ANTI-FLAG(R) M2 antibody produced in mouse 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4) | 50UG
Catalog No. NC1772686