Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti-Glucagon antibody produced in mouse, 100 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G2654100UL
Encompass
Monoclonal Anti-Glucagon antibody produced in mouse has been used: for the immunostaining of pancreatic tissue, in flow cytometry and immunofluorescence imaging of pancreas cells, and for immunohistochemistry and morphology of pancreas.
Catalog No. NC1844256