Learn More
ACROBiosystems Monoclonal Anti-Human IgG Antibody Europium chelate (for TR-FRET) is expressed from Mouse Hybridoma, 1000 tests
Catalog No. 5000807426
Encompass_Preferred
Monoclonal Anti-Human IgG Antibody Europium chelate (for TR-FRET) is expressed from Mouse Hybridoma
Catalog No. 50-008-07426 Supplier ACROBiosystems Supplier No. FRTTD021000TESTS
Monoclonal Anti-Human IgG Antibody Europium chelate (for TR-FRET) is expressed from Mouse Hybridoma