Learn More
ACROBiosystems Monoclonal Anti-Influenza A (H1N1) Hemagglutinin Antibody, Human IgG1, Clone 22F12, 1 mg (lyophilized)
Catalog No. NC3877608
Encompass_Preferred
Monoclonal antibody (clone 22F12) that specifically recognizes the hemagglutinin (HA) protein from Influenza A [Wisconsin/67/2022] (H1N1). Recombinant human IgG1, unconjugated, provided lyophilized and protein A/G purified. Validated by SEC-MALS and suitable for ELISA and western blot.
  • Recombinant human IgG1, kappa light chain.
  • Monoclonal clone 22F12.
  • Lyophilized formulation with trehalose stabilizer.
  • Protein A/G purified, ~90% purity by SEC-MALS.
  • Recommended for ELISA (0.05-50 ng/mL) and usable in western blot.
  • Available sizes: 100 μg and 1 mg (500 μg x 2).
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw cycles.
Catalog No. NC3877608 Supplier ACROBiosystems Supplier No. HA1MY21781MG
Monoclonal antibody (clone 22F12) that specifically recognizes the hemagglutinin (HA) protein from Influenza A [Wisconsin/67/2022] (H1N1). Recombinant human IgG1, unconjugated, provided lyophilized and protein A/G purified. Validated by SEC-MALS and suitable for ELISA and western blot.
  • Recombinant human IgG1, kappa light chain.
  • Monoclonal clone 22F12.
  • Lyophilized formulation with trehalose stabilizer.
  • Protein A/G purified, ~90% purity by SEC-MALS.
  • Recommended for ELISA (0.05-50 ng/mL) and usable in western blot.
  • Available sizes: 100 μg and 1 mg (500 μg x 2).
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw cycles.