Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-Influenza A [Wisconsin/67/2022] Neuraminidase (NA) Antibody, Human IgG1 (4A2), 100 μg

Catalog No. NC3845541
Encompass_Preferred

Chimeric monoclonal antibody against Influenza A (H1N1) neuraminidase (NA) from A/Wisconsin/67/2022, recombinantly expressed in HEK293 and supplied as an unconjugated, lyophilized product suitable for assay development and characterization.

  • Chimeric format with mouse variable regions and human IgG1 constant domains.
  • Host expressed in HEK293; variable region originally from mouse.
  • Isotype: human IgG1, kappa.
  • Target: Influenza A (H1N1) neuraminidase from A/Wisconsin/67/2022.
  • Size: 100 μg (also available as 1 mg, 500 μg x 2).
  • Purity: ~95% by SDS-PAGE; ~90% by SEC-MALS.
  • Formulation: lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Storage: lyophilized -20°C to -70°C for 12 months; reconstituted material stable at -70°C for 3 months.
  • Recommended application: ELISA (recommended range 0.06-125 ng/mL).
  • Conjugation: unconjugated.

Catalog No. NC3845541 Supplier ACROBiosystems Supplier No. NE1MY2159100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chimeric monoclonal antibody against Influenza A (H1N1) neuraminidase (NA) from A/Wisconsin/67/2022, recombinantly expressed in HEK293 and supplied as an unconjugated, lyophilized product suitable for assay development and characterization.

  • Chimeric format with mouse variable regions and human IgG1 constant domains.
  • Host expressed in HEK293; variable region originally from mouse.
  • Isotype: human IgG1, kappa.
  • Target: Influenza A (H1N1) neuraminidase from A/Wisconsin/67/2022.
  • Size: 100 μg (also available as 1 mg, 500 μg x 2).
  • Purity: ~95% by SDS-PAGE; ~90% by SEC-MALS.
  • Formulation: lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Storage: lyophilized -20°C to -70°C for 12 months; reconstituted material stable at -70°C for 3 months.
  • Recommended application: ELISA (recommended range 0.06-125 ng/mL).
  • Conjugation: unconjugated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.