Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti Tryptophan
Catalog No. 502906577
Encompass
The tryptophan hydroxylase 1 gene (TPH1) is a 444 amino acid protein encoded by the gene spanning 29 kbp with11 exons on human chromosome 11p15.3-p14. The enzyme is expressed in the gut spleen thymus pituitary and also in the pineal gland.
Catalog No. 50-290-6577 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T0678100UL
The tryptophan hydroxylase 1 gene (TPH1) is a 444 amino acid protein encoded by the gene spanning 29 kbp with11 exons on human chromosome 11p15.3-p14. The enzyme is expressed in the gut spleen thymus pituitary and also in the pineal gland.