Learn More
Cell Signaling Technology Cleaved PARP (Asp214) (D64E10) XP(R) Rabbit mAb (Alexa Fluor(R) 488 Conjugate) 100 ul
SDP
Supplier:  Cell Signaling Technology 9148S
Encompass
9148S , PARP, a 116 kDa nuclear poly (ADP-ribose) polymerase, appears to be involved in DNA repair in response to environmental stress (1). This protein can be cleaved by many ICE-like caspases in vitro (2,3) and is one of the main cleavage targets of caspase-3 in vivo (4,5). In human PARP, the cleavage occurs between Asp214 and Gly215, which separates the PARP amino-terminal DNA binding domain (24 kDa) from the carboxy-terminal catalytic domain (89 kDa) (2,4). PARP helps cells to maintain their viability; cleavage of PARP facilitates cellular disassembly and serves as a marker of cells undergoing apoptosis (6).
Catalog No. 50-190-509