Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-PEG Antibody, Human IgG1, Recombinant, 500 μg

Catalog No. NC3484278
Encompass_Preferred

This recombinant monoclonal antibody specifically recognizes polyethylene glycol (PEG), a synthetic polymer commonly used as an excipient and stabilizer in pharmaceutical therapeutics. It is composed of mouse variable regions genetically fused to human IgG1 constant domains and is recombinantly expressed in HEK293 cells. This antibody serves as a valuable tool for detecting PEGylated proteins and drugs in research and quality control applications, helping to assess drug efficacy and immunogenicity.

  • Host species: mouse
  • Isotype: human IgG1, kappa
  • Purification method: protein A purified
  • Formulation: lyophilized from a 0.22 μm-filtered solution in PBS (pH 3.0) containing HAC and trehalose
  • Recommended application: ELISA (0.2 to 2 μg/mL)
  • Storage: -20°C or lower in lyophilized state

Catalog No. NC3484278 Supplier ACROBiosystems Supplier No. PEGM731500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This recombinant monoclonal antibody specifically recognizes polyethylene glycol (PEG), a synthetic polymer commonly used as an excipient and stabilizer in pharmaceutical therapeutics. It is composed of mouse variable regions genetically fused to human IgG1 constant domains and is recombinantly expressed in HEK293 cells. This antibody serves as a valuable tool for detecting PEGylated proteins and drugs in research and quality control applications, helping to assess drug efficacy and immunogenicity.

  • Host species: mouse
  • Isotype: human IgG1, kappa
  • Purification method: protein A purified
  • Formulation: lyophilized from a 0.22 μm-filtered solution in PBS (pH 3.0) containing HAC and trehalose
  • Recommended application: ELISA (0.2 to 2 μg/mL)
  • Storage: -20°C or lower in lyophilized state

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.