Learn More
Analytical Sales & Services Monocon Gen II Single Heater, Touchscreen Interface, Up to 250 W Power, Each
Catalog No. NC2876269
Encompass
The MONOCON GEN II SINGLE HEATER is an advanced column heater sleeve controller designed for precise temperature management in scientific applications. It features a responsive touchscreen interface for easy operation, supports Gen II HotSleeve+ heaters and Peltier coolers, and offers a wide temperature range (5 ºC to 200 ºC) with a max heater power of 250 W. Its compact design and robust connectivity options make it versatile for various lab environments, ensuring efficient and reliable thermal control.
Catalog No. NC2876269 Supplier Analytical Sales & Services Supplier No. MONOCON
The MONOCON GEN II SINGLE HEATER is an advanced column heater sleeve controller designed for precise temperature management in scientific applications. It features a responsive touchscreen interface for easy operation, supports Gen II HotSleeve+ heaters and Peltier coolers, and offers a wide temperature range (5 ºC to 200 ºC) with a max heater power of 250 W. Its compact design and robust connectivity options make it versatile for various lab environments, ensuring efficient and reliable thermal control.