Learn More
TARGETMOL CHEMICALS INC Monocrotaline 200MG
SDP
Catalog No. 502878672
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Monocrotaline (Crotaline) is a pyrrolizidine alkaloid extracted from the seeds of the Crotalaria plant. Monocrotaline has certain cellular activity against HepG2 cells and can induce pulmonary hypertension in rodents. Purity 99.59%
Catalog No. 50-287-8672 Supplier TARGETMOL CHEMICALS INC Supplier No. T2803200MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Monocrotaline (Crotaline) is a pyrrolizidine alkaloid extracted from the seeds of the Crotalaria plant. Monocrotaline has certain cellular activity against HepG2 cells and can induce pulmonary hypertension in rodents. Purity 99.59%