Learn More
Medchemexpress LLC HY-N0750 50mg Medchemexpress, Monocrotaline CAS:315-22-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN075050MG
Encompass_Preferred
Medchemexpress, HY-N0750 50mg Monocrotaline CAS:315-22-0 Monocrotaline is an pyrrolizidine alkaloid extracted from the seeds of the Crotalaria spectabilis plant to induce pulmonary hypertension in rodents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3699