Learn More
ACCU-SCOPE™ Monocular Microscope with Built-in WiFi Camera
SDPSCI_ED
Catalog No. S00159 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Objective Types:
DIN Achromatic
Catalog No. Objective Types
S00159 DIN Achromatic
1 options
Catalog No. S00159 Supplier ACCU-SCOPE™ Supplier No. EXC101WM
Add to Cart
Add to Cart
The EXC-101-WM features a Binocular Microscope with built-in 2MP WiFi camera, 4x, 10x, 40x, and 100x oil immersion objectives, mechanical stage, rotatable tablet holder with Android tablet.
  • Monocular viewing head
  • Built-in 2MP WiFi camera, spring-loaded, rotatable tablet holder with Android tablet that attaches to top of viewing head, and rechargeable LED illumination
  • WF10x/18 mm eyepiece (paired - one w/pointer), diopter adjustment
  • Reversed quadruple nosepiece
  • DIN Achromat 4x, 10x, SP40xR and 100xR oil objectives
  • Double-layer Mechanical stage w/ low position coaxial controls
  • Abbe condenser, N.A. 1.25 with iris diaphragm and filter holder
  • Coaxial coarse and fine focusing controls
  • Variable LED illuminator, UL Listed
  • Dust cover, power cord and manual
Specifications
Eyepiece 10X/18 mm, with pointer
Head Style Monocular
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types DIN Achromatic
Product Type Digital Microscope System Package
Voltage 100/240 V