Learn More
Promocell GmbH PromoCell Monocyte Attachment Medium 250 ml, ready-to-use
Catalog No. 50306290
Encompass_Preferred
The Monocyte Attachment Medium allows for the efficient adherence selection of monocytes from freshly isolated human mononuclear cells while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes is dispensable prior to experiments requiring adherent monocytes as a starting material.With properly performed washing steps (see Instruction Manual) large numbers of 80-90% pure monocytes can be obtained with the PromoCell Monocyte Attachment Medium. Typical applications are generation of dendritic cells / macrophages as well as phagocytosis assays.
Catalog No. 50-306-290 Supplier Promocell GmbH Supplier No. C28051
The Monocyte Attachment Medium allows for the efficient adherence selection of monocytes from freshly isolated human mononuclear cells while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes is dispensable prior to experiments requiring adherent monocytes as a starting material.With properly performed washing steps (see Instruction Manual) large numbers of 80-90% pure monocytes can be obtained with the PromoCell Monocyte Attachment Medium. Typical applications are generation of dendritic cells / macrophages as well as phagocytosis assays.