Learn More
ATCC Monocytes derived from; DYS0100 Induced; Pluripotent Stem Cell; Human (Homo sapiens)
Catalog No. 502383755
Encompass
iPSC-derived Monocytes; DYS0100 are induced pluripotent stem cells that have been differentiated into CD14+ monocytes. These cells can be used in cancer immunology research, drug development, and toxicity screening.
Catalog No. 50-238-3755 Supplier ATCC Supplier No. ACS7030
iPSC-derived Monocytes; DYS0100 are induced pluripotent stem cells that have been differentiated into CD14+ monocytes. These cells can be used in cancer immunology research, drug development, and toxicity screening.