Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethanolamine Hydrochloride, Approx. 98%, Spectrum™ Chemical
SDP

Catalog No. 18604678 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-678 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-678 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1456100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1456, 2002-24-6, C2H7NO·HCl

Spectrum™ Chemical Monoethanolamine Hydrochloride , also known as ethanolamine or 2-aminoethanol, is an organic chemical compound used as feedstock in the production of emulsifiers, detergents, and pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
IUPAC Name hydrogen 2-aminoethan-1-ol chloride
SMILES [H+].[Cl-].NCCO

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity ∼98%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Monoethanolamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.