Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethanolamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018854 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8854 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8854 Supplier MilliporeSigma Supelco Supplier No. PHR14901ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1436°F
CAS 141-43-5
Vapor Pressure 0.2 mmHg (20°C)
Vapor Density 2.1 (vs. air)
Density 1.012 g/mL (at 25°C)
Flash Point 91°C - Pensky-Martens°Closed°Cup
Linear Formula NH2 CH2 CH2 OH
MDL Number MFCD00008183
Melting Point 10°C to 11°C
Boiling Point 170°C; 69°C to 70°C (10 mmHg)
Synonym Ethanolamine; 2-Aminoethanol; 2-Aminoethyl alcohol; ETA; MEA; MEA 90; MEA-LCI; Monoethanolamine
Type Certified Reference Material
Quantity 1 mL
Formula Weight 61.08
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.