Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethyl Adipate 97.0+%, TCI America™
SDP

Catalog No. A1223500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1223500G 500 g
A122325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1223500G Supplier TCI America Supplier No. A1223500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 626-86-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00004419
InChI Key UZNLHJCCGYKCIL-UHFFFAOYSA-N
Synonym adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester
PubChem CID 12295
IUPAC Name 6-ethoxy-6-oxohexanoic acid
SMILES CCOC(=O)CCCCC(=O)O

Specifications

Quantity 500 g
Formula Weight 174.20
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material Monoethyl Adipate
EINECSNumber (2)-0861
RTECSNumber AV2030000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.