Learn More
Medchemexpress LLC Monoethyl fumarate | 2459-05-4 | MFCD00002699 | 99.6% | 144.13 | C6H8O4 | 5 G
SDP
Catalog No. 5000300949
Encompass_Preferred
Monoethyl fumarate is the monoethyl ester of fumaric acid used primarily in research as an activator of the Nrf2 pathway and as a preservative and polymerization agent for macromolecular materials. It is provided as a high-purity solid suitable for biochemical and materials applications.
  • Activates the Nrf2 signaling pathway relevant for redox research.
  • High purity (≈99.6%), suitable for analytical and research use.
  • Solid form with defined storage stability under recommended conditions.
  • Useful as a preservative and polymerization agent for macromolecular materials.
  • Available in multiple pack sizes and solution formats for flexible use.
Catalog No. 50-003-00949 Supplier Medchemexpress LLC Supplier No. HYW0196965G
Monoethyl fumarate is the monoethyl ester of fumaric acid used primarily in research as an activator of the Nrf2 pathway and as a preservative and polymerization agent for macromolecular materials. It is provided as a high-purity solid suitable for biochemical and materials applications.
  • Activates the Nrf2 signaling pathway relevant for redox research.
  • High purity (≈99.6%), suitable for analytical and research use.
  • Solid form with defined storage stability under recommended conditions.
  • Useful as a preservative and polymerization agent for macromolecular materials.
  • Available in multiple pack sizes and solution formats for flexible use.