Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monoethyl fumarate | 2459-05-4 | MFCD00002699 | 99.6% | 144.13 g/mol | C6H8O4 | 100 G
SDP

Catalog No. 5000300971
Encompass_Preferred

Monoethyl fumarate is the monoethyl ester of fumaric acid, supplied as a powder for laboratory research. It functions as an NRF2 activator and is used as a preservative and polymerization agent in macromolecular material studies. For research use only; not for human consumption.

  • 99.6% purity.
  • Molecular weight 144.13 g/mol.
  • Molecular formula C6H8O4.
  • Off-white powder form.
  • Used as an NRF2 activator and polymerization preservative.
  • Store powder at -20°C for up to 3 years.

Catalog No. 50-003-00971 Supplier Medchemexpress LLC Supplier No. HYW019696100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoethyl fumarate is the monoethyl ester of fumaric acid, supplied as a powder for laboratory research. It functions as an NRF2 activator and is used as a preservative and polymerization agent in macromolecular material studies. For research use only; not for human consumption.

  • 99.6% purity.
  • Molecular weight 144.13 g/mol.
  • Molecular formula C6H8O4.
  • Off-white powder form.
  • Used as an NRF2 activator and polymerization preservative.
  • Store powder at -20°C for up to 3 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.