Learn More
Selleck Chemical LLC Monoethyl malonic acid S3366-100mg
SDP
Catalog No. 503108954
Encompass_Preferred
Monoethyl malonic acid (Ethyl hydrogen malonate Monoethyl malonate 3-Ethoxy-3-oxopropanoic acid) is the major urinary metabolites of ethyl 3-ethoxypropionate (EEP)
Catalog No. 50-310-8954 Supplier Selleck Chemical LLC Supplier No. S3366100MG
Monoethyl malonic acid (Ethyl hydrogen malonate Monoethyl malonate 3-Ethoxy-3-oxopropanoic acid) is the major urinary metabolites of ethyl 3-ethoxypropionate (EEP)