Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monoethyl phthalate | 1219806-03-7 | MFCD00462698 | 98.6% | 194.18 g/mol | C10H10O4 | 5 MG
SDP

Catalog No. 5000263059
Encompass_Preferred

Monoethyl phthalate is a high-purity analytical standard used as a phthalate metabolite reference for biochemical assays, method development, and mass spectrometry. It is supplied as a colorless to light-yellow liquid suitable for calibration, quantitation, and metabolite identification in research and analytical laboratories.

  • High purity (98.6%) suitable for analytical applications.
  • Molecular weight 194.18 g/mol for accurate mass calibration.
  • Chemical formula C10H10O4 for structural reference.
  • Available in small pack sizes (5 mg) for trace-level analysis.
  • Provided as a liquid with density 1.0583 g/cm3 for solution preparation.
  • Includes datasheet and certificate of analysis for batch traceability.

Catalog No. 50-002-63059 Supplier Medchemexpress LLC Supplier No. HY133668S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Monoethyl phthalate is a high-purity analytical standard used as a phthalate metabolite reference for biochemical assays, method development, and mass spectrometry. It is supplied as a colorless to light-yellow liquid suitable for calibration, quantitation, and metabolite identification in research and analytical laboratories.

  • High purity (98.6%) suitable for analytical applications.
  • Molecular weight 194.18 g/mol for accurate mass calibration.
  • Chemical formula C10H10O4 for structural reference.
  • Available in small pack sizes (5 mg) for trace-level analysis.
  • Provided as a liquid with density 1.0583 g/cm3 for solution preparation.
  • Includes datasheet and certificate of analysis for batch traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.