Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monoethyl phthalate | 2306-33-4 | MFCD00462698 | 98.6% | 194.18 g/mol | C10H10O4 | 5 MG
SDP

Catalog No. 5000262714
Encompass_Preferred

Monoethyl phthalate is a monoester metabolite of diethyl phthalate commonly used as an analytical reference standard in environmental, toxicology, and biomonitoring research. Supplied as a high-purity solid, it is intended for quantitative analysis, method development, and quality control in laboratories.

  • CAS number 2306-33-4
  • Molecular formula C10H10O4
  • Molecular weight 194.18 g/mol
  • Purity 98.6%
  • Package size 5 mg
  • Suitable for biomonitoring and exposure studies
  • Use as an analytical reference standard and for method development

Catalog No. 50-002-62714 Supplier Medchemexpress LLC Supplier No. HY1336685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoethyl phthalate is a monoester metabolite of diethyl phthalate commonly used as an analytical reference standard in environmental, toxicology, and biomonitoring research. Supplied as a high-purity solid, it is intended for quantitative analysis, method development, and quality control in laboratories.

  • CAS number 2306-33-4
  • Molecular formula C10H10O4
  • Molecular weight 194.18 g/mol
  • Purity 98.6%
  • Package size 5 mg
  • Suitable for biomonitoring and exposure studies
  • Use as an analytical reference standard and for method development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.