Learn More
Medchemexpress LLC Monoethyl phthalate | 2306-33-4 | MFCD00462698 | 98.6% | C10H10O4 | 10MG
SDP
Catalog No. 5000259064
Encompass_Preferred
Monoethyl phthalate is a metabolite of diethyl phthalate used as an analytical reference and research reagent to study phthalate exposure. It serves as a urinary biomarker in toxicology and epidemiology studies investigating associations with thyroid cancer and benign thyroid nodules. The material is supplied at high reported purity and is suitable for quantitative analytical assays.
  • High purity suitable for analytical assays.
  • Used as a urinary biomarker in exposure and epidemiological studies.
  • Compatible with HPLC and mass spectrometry methods.
  • Available as solid or DMSO solution formats for flexibility.
  • Provided with CAS and chemical identifiers for traceability.
Catalog No. 50-002-59064 Supplier Medchemexpress LLC Supplier No. HY13366810MG
Monoethyl phthalate is a metabolite of diethyl phthalate used as an analytical reference and research reagent to study phthalate exposure. It serves as a urinary biomarker in toxicology and epidemiology studies investigating associations with thyroid cancer and benign thyroid nodules. The material is supplied at high reported purity and is suitable for quantitative analytical assays.
  • High purity suitable for analytical assays.
  • Used as a urinary biomarker in exposure and epidemiological studies.
  • Compatible with HPLC and mass spectrometry methods.
  • Available as solid or DMSO solution formats for flexibility.
  • Provided with CAS and chemical identifiers for traceability.