Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monoethyl phthalate-d4 | 1219806-03-7 | 98.3% | C10H6D4O4 | 10MG
SDP

Catalog No. 5000260406
Encompass_Preferred

Monoethyl phthalate-d4 is a deuterium-labeled stable-isotope reference material used as an internal standard or tracer in analytical chemistry, particularly mass spectrometry and metabolite studies. It is supplied as a high-purity powder in small milligram quantities and is accompanied by datasheet and COA documentation; follow recommended storage conditions to maintain stability.

  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High purity (~98.3%).
  • Molecular formula C10H6D4O4; molecular weight 198.21 g/mol.
  • Supplied in small mg pack sizes suitable for analytical use.
  • Datasheet and certificate of analysis available for traceability.
  • Store powder at -20°C for long term; in solvent store at -80°C for up to 6 months.

Catalog No. 50-002-60406 Supplier Medchemexpress LLC Supplier No. HY133668S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoethyl phthalate-d4 is a deuterium-labeled stable-isotope reference material used as an internal standard or tracer in analytical chemistry, particularly mass spectrometry and metabolite studies. It is supplied as a high-purity powder in small milligram quantities and is accompanied by datasheet and COA documentation; follow recommended storage conditions to maintain stability.

  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High purity (~98.3%).
  • Molecular formula C10H6D4O4; molecular weight 198.21 g/mol.
  • Supplied in small mg pack sizes suitable for analytical use.
  • Datasheet and certificate of analysis available for traceability.
  • Store powder at -20°C for long term; in solvent store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.