Learn More
Medchemexpress LLC Monoethyl pimelate | 33018-91-6 | MFCD00014383 | >98.0% | 188.22 g/mol | C9H16O4 | 100 G
SDP
Catalog No. 5000303214
Encompass_Preferred
Monoethyl pimelate is the monoethyl ester of pimelic acid used as a chemical building block and PROTAC linker for small-molecule degrader synthesis. It serves as a synthetic intermediate in esterification and linker-construction chemistry and is supplied with product documentation for research use.
  • Monoethyl ester of pimelic acid.
  • Function as a PROTAC linker and synthetic intermediate.
  • Reported molecular formula C9H16O4 and molecular weight 188.22 g/mol.
  • Documentation includes a product data sheet, SDS, and handling instructions.
  • Available in multiple package sizes, including 100 g.
Catalog No. 50-003-03214 Supplier Medchemexpress LLC Supplier No. HYW016087100G
Monoethyl pimelate is the monoethyl ester of pimelic acid used as a chemical building block and PROTAC linker for small-molecule degrader synthesis. It serves as a synthetic intermediate in esterification and linker-construction chemistry and is supplied with product documentation for research use.
  • Monoethyl ester of pimelic acid.
  • Function as a PROTAC linker and synthetic intermediate.
  • Reported molecular formula C9H16O4 and molecular weight 188.22 g/mol.
  • Documentation includes a product data sheet, SDS, and handling instructions.
  • Available in multiple package sizes, including 100 g.