Learn More
Medchemexpress LLC Monoethyl pimelate | 33018-91-6 | MFCD00014383 | >98.0% | 188.22 g/mol | C9H16O4 | 25 G
SDP
Catalog No. 5000304195
Encompass_Preferred
Monoethyl pimelate is the monoethyl ester of pimelic acid used as a PROTAC linker and intermediate in small-molecule synthesis. It serves as an esterified building block for assembling linkers and other functionalized compounds in medicinal chemistry research.
  • Used as a PROTAC linker in bifunctional molecule design.
  • Suitable as an intermediate in organic synthesis.
  • Reported high purity, commonly >98%.
  • Molecular formula C9H16O4 and molecular weight 188.22 g/mol.
  • Provided with documentation to support research use.
Catalog No. 50-003-04195 Supplier Medchemexpress LLC Supplier No. 25GHYW016087
Monoethyl pimelate is the monoethyl ester of pimelic acid used as a PROTAC linker and intermediate in small-molecule synthesis. It serves as an esterified building block for assembling linkers and other functionalized compounds in medicinal chemistry research.
  • Used as a PROTAC linker in bifunctional molecule design.
  • Suitable as an intermediate in organic synthesis.
  • Reported high purity, commonly >98%.
  • Molecular formula C9H16O4 and molecular weight 188.22 g/mol.
  • Provided with documentation to support research use.