Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Monoethylene Glycol 3,000 ppm w/v Solution (1L)
SDP

Catalog No. NC0554161
Encompass_Preferred

Monoethylene Glycol 3,000 ppm w/v Solution (1L)

Catalog No. NC0554161 Supplier Aqua Solutions Supplier No. EQS511
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoethylene Glycol 3,000 ppm w/v Solution (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.