Learn More
McKesson General Medical McKesson Monofilament Sensory Test 10 Gram
Catalog No. NC1795879
Encompass
McKesson Monofilament Foot Screen Test. Monofilaments are a foot screening device used to assess loss of protective sensation.. Helps identify patients at risk of developing diabetic foot ulcers. Not Made with Natural Rubber Latex.
This item is not returnable. View return policy
Catalog No. NC1795879 Supplier McKesson General Medical Supplier No. 1074987
McKesson Monofilament Foot Screen Test. Monofilaments are a foot screening device used to assess loss of protective sensation.. Helps identify patients at risk of developing diabetic foot ulcers. Not Made with Natural Rubber Latex.