Learn More
Genscript Corporation Monofinity A Resin
Catalog No. 5000057853
Encompass
GenScript s Monofinity A Resin is made of recombinant protein A as ligand which not only keeps the specific binding capacity to Fc region of immunoglobulin molecules but also enhances chemical stability under Cleaning-in-place (CIP) conditions
Catalog No. 50-000-57853 Supplier Genscript Corporation Supplier No. L0043325
GenScript s Monofinity A Resin is made of recombinant protein A as ligand which not only keeps the specific binding capacity to Fc region of immunoglobulin molecules but also enhances chemical stability under Cleaning-in-place (CIP) conditions