Learn More
Genscript Corporation Monofinity A Resin Prepacked C
Catalog No. 5000057107
Encompass
GenScript s Monofinity A Resin is made of recombinant protein A as ligand which not only keeps the specific binding capacity to Fc region of immunoglobulin molecules but also enhances chemical stability under Cleaning-in-place (CIP) conditions Monofinity A Resin Prepacked Column 1mlx1
Catalog No. 50-000-57107 Supplier Genscript Corporation Supplier No. L0043311
GenScript s Monofinity A Resin is made of recombinant protein A as ligand which not only keeps the specific binding capacity to Fc region of immunoglobulin molecules but also enhances chemical stability under Cleaning-in-place (CIP) conditions Monofinity A Resin Prepacked Column 1mlx1