Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Corvac™ Silicone-Coated Tubes with Red and Gray Mottled Stoppers

Catalog No. 22029313
Change view
Click to view available options
Capacity (Metric):
4 mL
Tube Size:
13 x 75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Tube Size
22-029313 4 mL 13 x 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-029313 Supplier Covidien Supplier No. 8881302072

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Integrated serum separator tubes

Improved performance and more sample yield for laboratory procedures requiring serum

  • Contains a positive gel barrier between blood cells and serum
  • Interior silicone glass powder activates the clotting of blood
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging; tubes are sterilized by gamma radiation
  • Crown Corvac™ gel placement for improved performance and more sample yield
  • TDM compatible
  • Allows for sensitive and routine pharmacokinetic testing
  • Plastic coating prevents tube cracking
  • Can be used with both angle-head and horizontal-head centrifuges

  • Laboratory procedures requiring serum with a positive gel barrier between blood cells and serum; general chemistry, serology and toxicology procedures

Specifications

Product Type Integrated Serum Separator Tube
Sterility Sterile
Capacity (Metric) 4 mL
Closure Color Red/Gray
Width (English) 0.51 in.
Width (Metric) 13 mm
Height (English) 2.95 in.
Height (Metric) 75 mm
Tube Size 13 x 75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.