Learn More
Central Infusion Alliance Inc Monoject™ Tuberculin Syringe, 1 mL, Luer Lock Tip, Tip Cap
SDP
Catalog No. NC3974433
Encompass
The Monoject™ Tuberculin Syringe is a disposable plastic syringe with a 1 mL capacity, featuring a Luer Lock tip for secure connections. It includes a pharmacy grade tip cap and is designed with a clear barrel and large, bold graduations for accurate measurements in various laboratory and medical applications.
  • Equipped with a pharmacy grade tip cap.
  • Features large, bold graduations for clear visibility.
  • Designed with a crystal clear barrel for easy volume assessment.
  • Includes extended finger flanges for improved handling.
  • Provided sterile for safe use.
  • Constructed from durable plastic.
  • Designed without a safety feature.
Catalog No. NC3974433 Supplier Central Infusion Alliance Inc Supplier No. CIA1021106CS
The Monoject™ Tuberculin Syringe is a disposable plastic syringe with a 1 mL capacity, featuring a Luer Lock tip for secure connections. It includes a pharmacy grade tip cap and is designed with a clear barrel and large, bold graduations for accurate measurements in various laboratory and medical applications.
  • Equipped with a pharmacy grade tip cap.
  • Features large, bold graduations for clear visibility.
  • Designed with a crystal clear barrel for easy volume assessment.
  • Includes extended finger flanges for improved handling.
  • Provided sterile for safe use.
  • Constructed from durable plastic.
  • Designed without a safety feature.