Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl 2,6-Pyridinedicarboxylate 97.0+%, TCI America™
SDP

Catalog No. M25805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25805G 5 g
M25801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25805G Supplier TCI America Supplier No. M25805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7170-36-7 | C8H7NO4 | 181.15 g/mol

Chemical Identifiers

CAS 7170-36-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00806109
InChI Key VWIOMFMPIVMLIR-UHFFFAOYSA-N
Synonym 2,6-Pyridinedicarboxylic Acid Monomethyl Ester, Monomethyl Dipicolinate, Dipicolinic Acid Monomethyl Ester
PubChem CID 4019172
IUPAC Name 6-(methoxycarbonyl)pyridine-2-carboxylic acid
SMILES COC(=O)C1=CC=CC(=N1)C(O)=O

Specifications

Melting Point 153°C
Color White
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl 2,6-Pyridinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.