Learn More
Apexbio Technology LLC Monomethyl auristatin E, 10mg. Cas: 474645-27-7 MFCD: MFCD22124498. Antimitotic agent
SDP
Catalog No. 501904819
Encompass_Preferred
IC50: less than 1 nM for various cancer cell linesMonomethyl auristatin E(MMAE) is a potent antimitotic agent by blocking the polymerisation of tubulin. Microtubules play essential role in the function of the cell. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4819 Supplier Apexbio Technology LLC Supplier No. A363110
IC50: less than 1 nM for various cancer cell linesMonomethyl auristatin E(MMAE) is a potent antimitotic agent by blocking the polymerisation of tubulin. Microtubules play essential role in the function of the cell. Other sizes are available. Please inqury us for quote.