Learn More
Selleck Chemical LLC Monomethyl auristatin E (MMAE) 1mg 474645-27-7
SDP
Supplier:  Selleck Chemical LLC S77211MG
Encompass_Preferred
Monomethyl auristatin E (MMAE) is a synthetic antineoplastic agent, which is linked to a monoclonal antibody (MAB). It is also a microtubule-disrupting agent . *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6373