Learn More
Medchemexpress LLC Monomethyl auristatin E | 474645-27-7 | C39H67N5O7 | 25 MG
SDP
Catalog No. 5000374448
Encompass_Preferred
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10, acting as a potent mitotic inhibitor by preventing tubulin polymerization. It is widely used as a cytotoxic component in antibody-drug conjugates (ADCs) for treating various cancer types.
  • Potent mitotic inhibitor
  • Inhibits tubulin polymerization
  • Used in antibody-drug conjugates (ADCs)
  • Effective against various cancer types
Catalog No. 50-003-74448 Supplier Medchemexpress LLC Supplier No. 25MGHY15162R
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10, acting as a potent mitotic inhibitor by preventing tubulin polymerization. It is widely used as a cytotoxic component in antibody-drug conjugates (ADCs) for treating various cancer types.
  • Potent mitotic inhibitor
  • Inhibits tubulin polymerization
  • Used in antibody-drug conjugates (ADCs)
  • Effective against various cancer types