Learn More
Medchemexpress LLC Monomethyl auristatin E | 474645-27-7 | MFCD00868128 | C39H67N5O7 | 50 MG
SDP
Catalog No. 5000372897
Encompass_Preferred
Monomethyl auristatin E (MMAE) (Standard) is an analytical standard for research and analytical applications. This synthetic derivative of dolastatin 10 acts as a potent mitotic inhibitor by inhibiting tubulin polymerization and is widely used as a cytotoxic component in antibody-drug conjugates (ADCs) for various cancer types. This analytical standard is suitable for qualitative, quantitative, and methodological research experiments.
  • Analytical standard grade
  • Functions as a potent mitotic inhibitor
  • Inhibits tubulin polymerization
  • Utilized as a cytotoxic component in antibody-drug conjugates
  • Purity of 99.99%
  • Appears as a white to off-white solid
  • Commonly employed in HPLC, GC, and MS techniques
Catalog No. 50-003-72897 Supplier Medchemexpress LLC Supplier No. HY15162R50MG
Monomethyl auristatin E (MMAE) (Standard) is an analytical standard for research and analytical applications. This synthetic derivative of dolastatin 10 acts as a potent mitotic inhibitor by inhibiting tubulin polymerization and is widely used as a cytotoxic component in antibody-drug conjugates (ADCs) for various cancer types. This analytical standard is suitable for qualitative, quantitative, and methodological research experiments.
  • Analytical standard grade
  • Functions as a potent mitotic inhibitor
  • Inhibits tubulin polymerization
  • Utilized as a cytotoxic component in antibody-drug conjugates
  • Purity of 99.99%
  • Appears as a white to off-white solid
  • Commonly employed in HPLC, GC, and MS techniques