Learn More
Medchemexpress LLC Monomethyl auristatin E | 474645-27-7 | MFCD22124498 | 100.0% | 718.0 g/mol | C39H67N5O7 | 10 MG
SDP
Catalog No. 5000285921
Encompass_Preferred
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10 that functions as a highly potent antimitotic agent by inhibiting tubulin polymerization. Supplied as an analytical standard for laboratory research, it is used in assay development, impurity profiling, and synthesis workflows where a characterized reference material is required.
  • Potent antimitotic agent that inhibits tubulin polymerization.
  • Intended for research and analytical applications.
  • Useful as an analytical standard for assay development and quality control.
  • High purity reported on the product page (≈99.99%).
  • Supplied as a stable solid in small pack sizes for laboratory handling and storage.
Catalog No. 50-002-85921 Supplier Medchemexpress LLC Supplier No. HY15162R10MG
Monomethyl auristatin E (MMAE) is a synthetic derivative of dolastatin 10 that functions as a highly potent antimitotic agent by inhibiting tubulin polymerization. Supplied as an analytical standard for laboratory research, it is used in assay development, impurity profiling, and synthesis workflows where a characterized reference material is required.
  • Potent antimitotic agent that inhibits tubulin polymerization.
  • Intended for research and analytical applications.
  • Useful as an analytical standard for assay development and quality control.
  • High purity reported on the product page (≈99.99%).
  • Supplied as a stable solid in small pack sizes for laboratory handling and storage.