Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E | 474645-27-7 | MFCD22124498 | ≥98.0% | 1300.64 g/mol | C68H107N9O15 | 10 MG
SDP

Catalog No. 5000286474
Encompass_Preferred

Monomethyl auristatin E (MMAE) (GMP) is a GMP-grade cytotoxic agent that inhibits tubulin polymerization and is intended for research and development applications, including in vitro cytotoxicity assays and conjugation workflows. Supplied as a solid powder with high purity, it is suitable for laboratory workflows that require GMP-grade material.

  • High purity ≥98% (HPLC).
  • Potent tubulin polymerization inhibitor with low-nanomolar cellular activity.
  • Supplied as a solid powder suitable for conjugation workflows and assays.
  • Storage: powder -80°C for long term, -20°C for short term.
  • Available in small vial sizes appropriate for R&D use.
  • CAS number 474645-27-7 provided for unambiguous identification.

Catalog No. 50-002-86474 Supplier Medchemexpress LLC Supplier No. HY15162G10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl auristatin E (MMAE) (GMP) is a GMP-grade cytotoxic agent that inhibits tubulin polymerization and is intended for research and development applications, including in vitro cytotoxicity assays and conjugation workflows. Supplied as a solid powder with high purity, it is suitable for laboratory workflows that require GMP-grade material.

  • High purity ≥98% (HPLC).
  • Potent tubulin polymerization inhibitor with low-nanomolar cellular activity.
  • Supplied as a solid powder suitable for conjugation workflows and assays.
  • Storage: powder -80°C for long term, -20°C for short term.
  • Available in small vial sizes appropriate for R&D use.
  • CAS number 474645-27-7 provided for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.