Learn More
Medchemexpress LLC Monomethyl auristatin E-d8 | 2070009-72-0 | C₃₉H₅₉D₈N₅O₇ | 5 MG
SDP
Catalog No. 5000373220
Encompass_Preferred
Monomethyl auristatin E-d8 (MMAE-d8) is a deuterated labeled monomethyl auristatin E, functioning as a potent mitotic inhibitor and tubulin inhibitor. It is utilized in antibody-drug conjugates (ADC) as a cytotoxic payload and can serve as a tracer or internal standard for quantitative analysis.
  • Potent mitotic and tubulin inhibitor
  • Used in antibody-drug conjugates (ADC) as a cytotoxic payload
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets microtubule/tubulin and ADC cytotoxins
  • Relevant to cell cycle/DNA damage, cytoskeleton, and antibody-drug conjugate related research
Catalog No. 50-003-73220 Supplier Medchemexpress LLC Supplier No. HY15162A5MG
Monomethyl auristatin E-d8 (MMAE-d8) is a deuterated labeled monomethyl auristatin E, functioning as a potent mitotic inhibitor and tubulin inhibitor. It is utilized in antibody-drug conjugates (ADC) as a cytotoxic payload and can serve as a tracer or internal standard for quantitative analysis.
  • Potent mitotic and tubulin inhibitor
  • Used in antibody-drug conjugates (ADC) as a cytotoxic payload
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets microtubule/tubulin and ADC cytotoxins
  • Relevant to cell cycle/DNA damage, cytoskeleton, and antibody-drug conjugate related research